Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gogo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140943.3 | NP_649200.2 | 5 - 6 | 2409 | 64 | |
GenBank mRNA | BT009982.1 | - | 5 - 6 | 2407 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |