Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vir-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259075.2 | NP_001246004.2 | 3 - 4 | 461 | 79 | |
NM_001273483.1 | NP_001260412.1 | 3 - 4 | 461 | 79 | ||
NM_165010.3 | NP_723746.1 | 2 - 3 | 388 | 79 | ||
NM_165011.3 | NP_723747.1 | 3 - 4 | 472 | 79 | ||
NM_205966.2 | NP_995688.1 | 2 - 3 | 388 | 79 | ||
GenBank mRNA | AY094667.1 | - | 2 - 3 | 539 | 79 | |
BT044312.1 | - | 3 - 4 | 468 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |