Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yuri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038821.2 | NP_001033910.2 | 1 - 2 | 427 | 80 | |
NM_001038822.3 | NP_001033911.3 | 2 - 3 | 341 | 80 | ||
NM_001316397.1 | NP_001303326.1 | 5 - 6 | 1323 | 80 | ||
NM_165106.5 | NP_723903.3 | 4 - 5 | 1201 | 80 | ||
NM_165107.3 | NP_723904.1 | 4 - 5 | 1666 | 80 | ||
NM_165109.3 | NP_723906.1 | 5 - 6 | 1611 | 80 | ||
GenBank mRNA | BT001564.1 | - | 4 - 5 | 1664 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |