Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yuri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038822.3 | NP_001033911.3 | 1 - 2 | 93 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |