Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258491.3 | NP_001245420.1 | 13 - 14 | 4842 | 60 | |
NM_079882.4 | NP_524621.2 | 12 - 13 | 4816 | 60 | ||
NM_166756.3 | NP_726566.1 | 12 - 13 | 4512 | 60 | ||
NM_166757.3 | NP_726567.2 | 12 - 13 | 4660 | 60 | ||
NM_205873.3 | NP_995595.1 | 12 - 13 | 4512 | 60 | ||
GenBank mRNA | AJ320236.1 | - | 11 - 12 | 4469 | 60 | |
BT015268.1 | - | 12 - 13 | 4777 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |