Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258491.3 | NP_001245420.1 | 9 - 10 | 3812 | 80 | |
NM_079882.4 | NP_524621.2 | 8 - 9 | 3786 | 80 | ||
NM_166756.3 | NP_726566.1 | 8 - 9 | 3482 | 80 | ||
NM_166757.3 | NP_726567.2 | 8 - 9 | 3630 | 80 | ||
NM_205873.3 | NP_995595.1 | 8 - 9 | 3482 | 80 | ||
GenBank mRNA | AJ320236.1 | - | 7 - 8 | 3439 | 80 | |
BT015268.1 | - | 8 - 9 | 3747 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |