Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr36c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165207.1 | NP_724040.1 | 1 - 2 | 265 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Drosophila melanogaster |