Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ninaD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165237.2 | NP_724087.1 | 4 - 5 | 1371 | 106 | |
GenBank mRNA | BT031187.1 | - | 4 - 5 | 1369 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |