Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169618.2 | NP_001163089.2 | 4 - 5 | 1463 | 75 | |
NM_165611.3 | NP_724690.1 | 3 - 4 | 1364 | 75 | ||
GenBank mRNA | AF353512.1 | - | 3 - 4 | 1312 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |