Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stmA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014513.2 | NP_001014513.1 | 1 - 2 | 515 | 62 | |
NM_165624.2 | NP_724710.1 | 1 - 2 | 515 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |