Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wun | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259294.2 | NP_001246223.1 | 4 - 5 | 1048 | 80 | |
NM_057845.5 | NP_477193.1 | 3 - 4 | 648 | 80 | ||
NM_165674.3 | NP_724787.1 | 4 - 5 | 1048 | 80 | ||
GenBank mRNA | AF145595.1 | - | 3 - 4 | 632 | 80 | |
BT001729.1 | - | 4 - 5 | 1045 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |