Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103491.1 | NP_001096961.1 | 3 - 4 | 890 | 56 | |
NM_001298217.1 | NP_001285146.1 | 4 - 5 | 1270 | 56 | ||
NM_078581.3 | NP_511136.2 | 5 - 6 | 1650 | 56 | ||
GenBank mRNA | BT003214.1 | - | 5 - 6 | 1647 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |