Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ten-a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144717.4 | NP_001138189.1 | 17 - 18 | 6635 | 65 | |
NM_001144718.3 | NP_001138190.1 | 18 - 19 | 7340 | 65 | ||
NM_001169258.2 | NP_001162729.1 | 12 - 13 | 4569 | 65 | ||
NM_001169259.3 | NP_001162730.2 | 18 - 19 | 6745 | 65 | ||
NM_001169260.3 | NP_001162731.1 | 19 - 20 | 7611 | 65 | ||
NM_001169261.3 | NP_001162732.2 | 18 - 19 | 6662 | 65 | ||
NM_001258711.2 | NP_001245640.1 | 15 - 16 | 5584 | 65 | ||
NM_001258712.2 | NP_001245641.1 | 16 - 17 | 5617 | 65 | ||
NM_001258713.2 | NP_001245642.1 | 16 - 17 | 5629 | 65 | ||
NM_001272554.2 | NP_001259483.1 | 15 - 16 | 5484 | 65 | ||
NM_078582.4 | NP_511137.3 | 13 - 14 | 4770 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |