Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169275.2 | NP_001162746.1 | 1 - 2 | 852 | 75 | |
NM_001272597.2 | NP_001259526.2 | 1 - 2 | 852 | 75 | ||
NM_078592.3 | NP_511147.2 | 1 - 2 | 852 | 75 | ||
GenBank mRNA | AF195496.1 | - | 1 - 2 | 486 | 75 | |
BT015267.1 | - | 1 - 2 | 486 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |