Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103504.2 | NP_001096974.1 | 4 - 5 | 1793 | 100 | |
NM_078592.3 | NP_511147.2 | 4 - 5 | 2012 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |