Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NetA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258738.2 | NP_001245667.1 | 1 - 2 | 1695 | 56 | |
NM_001298314.1 | NP_001285243.1 | 1 - 2 | 1789 | 56 | ||
NM_078599.5 | NP_511154.3 | 1 - 2 | 1695 | 56 | ||
GenBank mRNA | AY089580.1 | - | 1 - 2 | 1692 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |