Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hiw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298329.1 | NP_001285258.1 | 21 - 22 | 15361 | 98 | |
NM_078604.4 | NP_511159.3 | 21 - 22 | 15361 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |