Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kat80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298388.1 | NP_001285317.1 | 5 - 6 | 1212 | 65 | |
NM_078639.3 | NP_523363.2 | 6 - 7 | 1113 | 65 | ||
NM_167488.2 | NP_727940.1 | 5 - 6 | 1212 | 65 | ||
GenBank mRNA | BT044203.1 | - | 5 - 6 | 1193 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |