Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpS5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298431.1 | NP_001285360.1 | 2 - 3 | 611 | 116 | |
NM_001298432.1 | NP_001285361.1 | 2 - 3 | 980 | 116 | ||
NM_001298433.1 | NP_001285362.1 | 1 - 2 | 409 | 116 | ||
NM_078658.3 | NP_523382.1 | 2 - 3 | 464 | 116 | ||
GenBank mRNA | AY075368.1 | - | 2 - 3 | 454 | 116 | |
U48394.1 | - | 2 - 3 | 458 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |