Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078719.4 | NP_523443.1 | 5 - 6 | 1955 | 88 | |
GenBank mRNA | BT053691.1 | - | 5 - 6 | 1927 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Drosophila melanogaster |