Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272996.1 | NP_001259925.1 | 2 - 3 | 556 | 87 | |
NM_001272997.1 | NP_001259926.1 | 2 - 3 | 547 | 87 | ||
NM_001272998.1 | NP_001259927.1 | 2 - 3 | 546 | 87 | ||
NM_078733.4 | NP_523457.1 | 2 - 3 | 529 | 87 | ||
NM_164475.2 | NP_722795.1 | 2 - 3 | 536 | 87 | ||
NM_164476.2 | NP_722796.1 | 2 - 3 | 642 | 87 | ||
NM_164477.3 | NP_722797.1 | 2 - 3 | 838 | 87 | ||
NM_164478.2 | NP_722798.1 | 2 - 3 | 510 | 87 | ||
NM_164479.2 | NP_722799.1 | 2 - 3 | 494 | 87 | ||
GenBank mRNA | AB035353.1 | - | 2 - 3 | 544 | 87 | |
AB035671.1 | - | 2 - 3 | 515 | 87 | ||
AY060343.1 | - | 2 - 3 | 814 | 87 | ||
AY081180.1 | - | 1 - 2 | 414 | 87 | ||
AY081181.1 | - | 2 - 3 | 498 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |