Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078783.3 | NP_523507.2 | 5 - 6 | 1699 | 99 | |
GenBank mRNA | AY069050.1 | - | 5 - 6 | 1665 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |