Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CdGAPr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169546.2 | NP_001163017.1 | 3 - 4 | 1803 | 90 | |
NM_001169547.2 | NP_001163018.1 | 3 - 4 | 1803 | 90 | ||
NM_001273671.2 | NP_001260600.1 | 4 - 5 | 1875 | 90 | ||
NM_136158.5 | NP_610002.1 | 3 - 4 | 1803 | 90 | ||
GenBank mRNA | AF218776.1 | - | 3 - 4 | 1792 | 90 | |
BT132944.1 | - | 3 - 4 | 1773 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |