Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299173.1 | NP_001286102.1 | 4 - 5 | 996 | 67 | |
NM_136189.4 | NP_610033.1 | 4 - 5 | 963 | 67 | ||
GenBank mRNA | AY060391.1 | - | 4 - 5 | 939 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |