Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipped-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103722.3 | NP_001097192.2 | 17 - 18 | 8981 | 99 | |
NM_001316406.1 | NP_001303335.1 | 16 - 17 | 8834 | 99 | ||
GenBank mRNA | DQ352451.1 | - | 17 - 18 | 9007 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |