Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadd45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136420.4 | NP_610264.1 | 1 | 810 | 71 | |
GenBank mRNA | BT088861.1 | - | 1 | 807 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |