Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nito | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136495.3 | NP_610339.1 | 3 - 4 | 2647 | 130 | |
GenBank mRNA | AF145664.1 | - | 3 - 4 | 2624 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |