Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079058.3 | NP_523782.1 | 2 - 3 | 1907 | 82 | |
NM_166289.2 | NP_725794.2 | 2 - 3 | 1907 | 82 | ||
GenBank mRNA | AF239722.1 | - | 2 - 3 | 1825 | 82 | |
BT003216.1 | - | 2 - 3 | 1915 | 82 | ||
BT125963.1 | - | 3 - 4 | 1727 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |