Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ord | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299831.1 | NP_001286760.1 | 2 - 3 | 703 | 69 | |
NM_166589.2 | NP_726303.1 | 2 - 3 | 2440 | 69 | ||
GenBank mRNA | BT122096.2 | - | 2 - 3 | 678 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |