Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078854.2 | NP_523578.1 | 7 - 8 | 2024 | 97 | |
GenBank mRNA | AJ269538.1 | - | 7 - 8 | 2024 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |