Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201901.2 | NP_001188830.1 | 2 - 3 | 776 | 90 | |
NM_001201902.1 | NP_001188831.1 | 1 - 2 | 670 | 90 | ||
NM_001299088.1 | NP_001286017.1 | 2 - 3 | 994 | 90 | ||
NM_078868.3 | NP_523592.1 | 1 - 2 | 1408 | 90 | ||
GenBank mRNA | BT004905.1 | - | 1 - 2 | 670 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |