Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tep4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273660.1 | NP_001260589.1 | 3 - 4 | 990 | 62 | |
NM_001273661.1 | NP_001260590.1 | 2 - 3 | 967 | 62 | ||
NM_001273662.1 | NP_001260591.1 | 3 - 4 | 990 | 62 | ||
NM_078879.3 | NP_523603.2 | 2 - 3 | 967 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |