Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Ek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259232.1 | NP_001246161.1 | 4 - 5 | 950 | 67 | |
NM_078913.4 | NP_523637.1 | 4 - 5 | 696 | 67 | ||
GenBank mRNA | BT088928.1 | - | 4 - 5 | 950 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |