Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Eo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078917.3 | NP_523641.2 | 2 - 3 | 391 | 65 | |
GenBank mRNA | AY070986.1 | - | 2 - 3 | 387 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |