Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Er | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078920.3 | NP_523644.1 | 1 - 2 | 152 | 114 | |
GenBank mRNA | BT025888.1 | - | 1 - 2 | 135 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |