Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or47a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078965.3 | NP_523689.1 | 2 - 3 | 499 | 84 | |
GenBank mRNA | AF156880.1 | - | 2 - 3 | 507 | 84 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |