Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tou | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103800.3 | NP_001097270.1 | 2 - 3 | 645 | 64 | |
NM_001273969.1 | NP_001260898.1 | 2 - 3 | 346 | 64 | ||
NM_001273970.1 | NP_001260899.1 | 3 - 4 | 611 | 64 | ||
NM_078977.5 | NP_523701.3 | 2 - 3 | 645 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |