Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078999.4 | NP_523723.1 | 6 - 7 | 2693 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |