Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4p2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136628.3 | NP_610472.1 | 6 - 7 | 1123 | 116 | |
GenBank mRNA | AY051564.1 | - | 6 - 7 | 1105 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |