Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144152.2 | NP_001137624.1 | 16 - 17 | 5485 | 64 | |
NM_001144153.2 | NP_001137625.1 | 15 - 16 | 5368 | 64 | ||
NM_001144154.2 | NP_001137626.1 | 13 - 14 | 4647 | 64 | ||
NM_001169627.2 | NP_001163098.1 | 15 - 16 | 5365 | 64 | ||
NM_001299320.1 | NP_001286249.1 | 14 - 15 | 4093 | 64 | ||
NM_001299321.1 | NP_001286250.1 | 15 - 16 | 5365 | 64 | ||
NM_136647.5 | NP_610491.4 | 14 - 15 | 4764 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |