Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144165.2 | NP_001137637.1 | 1 - 2 | 6073 | 79 | |
NM_001273930.1 | NP_001260859.1 | 1 - 2 | 3187 | 79 | ||
NM_136727.4 | NP_610571.2 | 1 - 2 | 6073 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |