Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038850.2 | NP_001033939.2 | 6 | 1024 | 81 | |
NM_001043071.2 | NP_001036536.1 | 6 | 789 | 81 | ||
NM_001316415.1 | NP_001303344.1 | 5 | 1568 | 81 | ||
GenBank mRNA | KM360087.1 | - | 6 | 784 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |