Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wash | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299405.1 | NP_001286334.1 | 2 | 1740 | 117 | |
NM_136895.4 | NP_610739.1 | 1 | 1695 | 117 | ||
GenBank mRNA | AY118440.1 | - | 1 | 1678 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |