Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eIF2Bdelta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169794.2 | NP_001163265.1 | 3 - 4 | 1339 | 83 | |
NM_137946.4 | NP_611790.1 | 4 - 5 | 1312 | 83 | ||
NM_166601.2 | NP_726320.1 | 2 - 3 | 1275 | 83 | ||
NM_166602.3 | NP_726321.1 | 3 - 4 | 1274 | 83 | ||
GenBank mRNA | AY069341.1 | - | 4 - 5 | 1290 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster Inventoried |