Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169803.2 | NP_001163274.1 | 6 - 7 | 1181 | 58 | |
NM_001169804.2 | NP_001163275.2 | 2 - 3 | 1341 | 58 | ||
NM_001169805.2 | NP_001163276.1 | 5 - 6 | 1008 | 58 | ||
NM_001169807.2 | NP_001163278.2 | 5 - 6 | 1294 | 58 | ||
NM_137970.5 | NP_611814.3 | 6 - 7 | 1039 | 58 | ||
NM_166616.3 | NP_726350.1 | 5 - 6 | 1294 | 58 | ||
NM_166617.2 | NP_726351.1 | 3 - 4 | 428 | 58 | ||
NM_166618.3 | NP_726352.1 | 4 - 5 | 777 | 58 | ||
GenBank mRNA | BT021326.1 | - | 6 - 7 | 1035 | 58 | |
BT133111.1 | - | 4 - 5 | 777 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |