Search
Search

Pym| Gene Symbol: Pym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166627.3 | NP_726372.1 | 1 | 839 | 147 | |
| GenBank mRNA | AF293388.1 | - | 1 | 804 | 147 | |
| AY070957.1 | - | 1 | 837 | 147 | ||
| AY071704.1 | - | 1 | 838 | 147 | ||
Set Membership: |
|