Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-Catr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143816.3 | NP_652073.2 | 2 - 3 | 556 | 76 | |
NM_166641.2 | NP_726389.4 | 2 - 3 | 556 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |