Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144282.2 | NP_001137754.1 | 13 - 14 | 3458 | 81 | |
NM_001299914.1 | NP_001286843.1 | 13 - 14 | 3458 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |