Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014552.3 | NP_001014552.1 | 8 - 9 | 3739 | 90 | |
NM_001014553.3 | NP_001014553.1 | 8 - 9 | 3675 | 90 | ||
NM_001202079.1 | NP_001189008.1 | 8 - 9 | 3702 | 90 | ||
NM_001202080.2 | NP_001189009.1 | 9 - 10 | 3699 | 90 | ||
NM_001202081.1 | NP_001189010.1 | 8 - 9 | 3696 | 90 | ||
NM_001202082.2 | NP_001189011.1 | 9 - 10 | 3699 | 90 | ||
NM_001259587.2 | NP_001246516.1 | 9 - 10 | 3763 | 90 | ||
NM_079136.4 | NP_523860.2 | 9 - 10 | 3859 | 90 | ||
NM_166704.4 | NP_726506.1 | 9 - 10 | 3795 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |