Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01821180_g1
See other TROL GE Assays ›
Gene Symbol:
trol
Gene Name:
terribly reduced optic lobes
Gene Aliases:
BcDNA:GM02481, CG12497, CG33675, CG33950, CG7981, CT23996, Dmel\CG33950, Dmel_CG33950, EG:BACR25B3.1, EG:BACR25B3.10, EG:BACR25B3.11, EG:BACR25B3.2, GC7891, MRE7, Pcan, Pcn, TROL, Trol, Trol-A, Trol-B, Troll, ZW-1, anon-WO0153538.72, l(1)3Ac, l(1)9-96, l(1)G0019, l(1)G0021, l(1)G0023, l(1)G0181, l(1)G0211, l(1)G0271, l(1)G0374, l(1)G0412, l(1)VA51, l(1)trol, l(1)zw1, l(1)zwl, pcan, troll, zw-1, zw1
Chromosome Location:
Chr.X: 2470460 - 2545427 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0284408
Assay ID
Dm01821180_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: trol
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001031862.4
NP_001027033.3
22 - 23
7777
57
NM_001031863.5
NP_001027034.2
29 - 30
8448
57
NM_001031864.4
NP_001027035.3
21 - 22
7417
57
NM_001031865.4
NP_001027036.3
22 - 23
7777
57
NM_001031866.4
NP_001027037.3
24 - 25
8206
57
NM_001031867.4
NP_001027038.3
31 - 32
9109
57
NM_001103399.4
NP_001096869.3
21 - 22
7723
57
NM_001169177.4
NP_001162648.3
30 - 31
8989
57
NM_001258562.3
NP_001245491.2
31 - 32
9109
57
NM_001258563.3
NP_001245492.2
18 - 19
6940
57
NM_001258564.3
NP_001245493.2
30 - 31
9058
57
NM_001258565.3
NP_001245494.2
20 - 21
7255
57
NM_001258566.4
NP_001245495.2
29 - 30
8291
57
NM_001258567.3
NP_001245496.2
19 - 20
7030
57
NM_001258568.3
NP_001245497.2
21 - 22
7657
57
NM_001258569.3
NP_001245498.2
19 - 20
7174
57
NM_001258570.3
NP_001245499.2
27 - 28
8653
57
NM_001258571.3
NP_001245500.2
24 - 25
8206
57
NM_001297893.2
NP_001284822.1
19 - 20
7171
57
NM_001297894.2
NP_001284823.1
21 - 22
7675
57
NM_001297895.2
NP_001284824.1
19 - 20
7021
57
NM_001297896.2
NP_001284825.1
25 - 26
8365
57
NM_001382068.1
NP_001368970.1
19 - 20
7030
57
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Drosophila melanogaster
Gene Ontology Categories:
Function(s)
positive regulation of neuroblast proliferation
defasciculation of motor neuron axon
motor neuron axon guidance
organ morphogenesis
tissue development
negative regulation of fibroblast growth factor receptor signaling pathway
maintenance of epithelial cell apical/basal polarity
positive regulation of fibroblast growth factor receptor signaling pathway
positive regulation of smoothened signaling pathway
response to anesthetic
positive regulation of semaphorin-plexin signaling pathway
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0